Ethyl 1-amino-1H-imidazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A845032-1G A845032-100MG A845032-250MG
103841-216EA 491 USD
103841-216 76845-460 77019-654 77019-656
Ethyl 1-amino-1H-imidazole-5-carboxylate 95%
Ethyl 1-amino-1H-imidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12755702
CAS Number: 1179361-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR