Ethyl-1,5-Dimethylpyrazole-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1,5-dimethyl-1H-pyrazole-3-carboxylate, 1,5-Dimethylpyrazole-3-carboxylic Acid Ethyl Ester

A202051-100G A202051-10G A202051-25G A202051-5G A202051-1G
103817-350EA 12 USD
103817-350 103847-986 76770-176 76770-178 77086-078 77086-080 77302-786
Ethyl-1,5-Dimethylpyrazole-3-carboxylate 98%
Ethyl-1,5-Dimethylpyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.20 g/mol
Boiling Pt: 258.8 °C (760 mm Hg)
Melting Pt: 39…41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085071
CAS Number: 5744-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR