Ethyl 1,4-dioxaspiro[4.5]decane-8-carboxylate 95%

Supplier: Ambeed
Warning

A214539-100G A214539-10G A214539-1G A214539-25G A214539-5G
103818-412EA 7 USD
103818-412 103849-116 76873-778 76873-780 77087-640 77087-642 77087-644
Ethyl 1,4-dioxaspiro[4.5]decane-8-carboxylate 95%
Ethyl 1,4-dioxaspiro[4.5]decane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₄
Storage Temperature: Ambient
MDL Number: MFCD00205582
CAS Number: 1489-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR