4-(1-Ethoxycarbonylcyclopropyl)phenylboronic acid, pinacol ester 95%

Supplier: Ambeed
Warning

A366373-250MG A366373-1G
103826-394EA 33 USD
103826-394 76817-440 76840-648
4-(1-Ethoxycarbonylcyclopropyl)phenylboronic acid, pinacol ester 95%
4-(1-Ethoxycarbonylcyclopropyl)phenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₅BO₄
Boiling Pt: 400.6±38.0 °C (760 mm Hg)
Flash Pt: 196.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18383419
CAS Number: 1257213-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR