Ethyl-1-((2'-cyano[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate

A164996-1G A164996-25G A164996-5G
103805-928EA 10 USD
103805-928 103844-186 77300-744 77300-746 77300-748
Ethyl-1-((2'-cyano[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate 95%
Ethyl-1-((2'-cyano[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate

Precaución: for Research Use Only

Formula: C₂₆H₂₃N₃O₃
MW: 425.4800 g/mol
Boiling Pt: 637.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09030636
CAS Number: 139481-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR