Ethyl (R)-(-)-4-cyano-3-hydroxybutyate 97%

Supplier: Ambeed
Warning

Synonyms: (R)-4-Cyano-3-hydroxybutyric acid ethyl ester, BUTANOIC ACID, 4-CYANO-3-HYDROXY-, ETHYL ESTER, (3R)-, Ethyl (R)-4-cyano-3-hydroxybutanoate, (R)-ethyl 4-cyano-3-hydroxybutanoate

A183042-25G A183042-100G A183042-10G A183042-500G
103846-156EA 5 USD
103846-156 77083-746 77111-474 77111-476 77111-478 77301-808
Ethyl (R)-(-)-4-cyano-3-hydroxybutyate 97%
Ethyl (R)-(-)-4-cyano-3-hydroxybutyate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
Boiling Pt: 270 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270839
CAS Number: 141942-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR