Ethyl (2-chlorobenzoyl)acetate 95%

Supplier: Ambeed
Warning

A422768-1G A422768-25G A422768-5G A422768-100G A422768-10G
103827-428EA 19 USD
103827-428 103838-476 76842-104 76842-106 76842-108 77016-480 77016-482
Ethyl (2-chlorobenzoyl)acetate 95%
Ethyl (2-chlorobenzoyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClO₃
Boiling Pt: 280.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03424811
CAS Number: 19112-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR