1,1-Diphenylethylene 98% stabilized
Supplier: Ambeed
|
Warning
|
Synonyms:
asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
Stabilization:
Stabilised with hydroquinone (HQ)
A104720-100G
A104720-10G
A104720-25G
A104720-500G
A104720-5G
103822-488EA
7
USD
103822-488
77160-412
77160-414
77160-416
77160-418
77283-262
1,1-Diphenylethylene 98% stabilized
1,1-Diphenylethylene
Precaución: for Research Use Only
Formula:
(C₆H₅)₂CCH₂ MW: 180.25 g/mol Boiling Pt: 270…271 °C (760 mm Hg) Melting Pt: 6 °C Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008583 CAS Number: 530-48-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...