Esculetin 98%

Supplier: Ambeed
Warning

Synonyms: 6,7-Dihydroxycoumarin, 6,7-Dihydroxy-2H-chromen-2-one

A155662-100G A155662-10G A155662-250MG A155662-1G A155662-25G A155662-5G
103804-788EA 19 USD
103804-788 103842-990 76997-518 76997-520 76997-522 77318-710 77318-712 77318-714
Esculetin 98%
Esculetin

Precaución: for Research Use Only

Formula: C₉H₆O₄
MW: 178.14 g/mol
Boiling Pt: 469.7 °C (760 mm Hg)
Melting Pt: 271…273 °C
Flash Pt: 201.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00006874
CAS Number: 305-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR