(R)-(-)-2-Amino-3-methyl-1-butanol 98%

Supplier: Ambeed
Warning

Synonyms: D(-)-2-Amino-3-methyl-1-butanol, (R)-(-)-2-Amino-3-methylbutanol, D-Valinol, D(-)-Valinol

A220193-100G A220193-10G A220193-1G A220193-25G A220193-5G A220193-500G
103818-842EA 6 USD
103818-842 103849-724 77288-212 77288-214 77303-538 77303-540 77303-542 77748-351
(R)-(-)-2-Amino-3-methyl-1-butanol 98%
(R)-(-)-2-Amino-3-methyl-1-butanol

Precaución: for Research Use Only

Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 186.8 °C (760 mm Hg)
Melting Pt: 35…36 °C
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064297
CAS Number: 4276-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR