D-(+)-Trehalose dihydrate 97%

Supplier: Ambeed
Warning

Synonyms: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate

A308914-100G A308914-10G A308914-500G A308914-1000G A308914-25G
103810-052EA 117 USD
103810-052 76816-874 76816-876 76816-878 77306-000 77306-002
D-(+)-Trehalose dihydrate 97%
D-(+)-Trehalose dihydrate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₁₁·2H₂O
MW: 378.33 g/mol
Boiling Pt: 675.4 °C (760 mm Hg)
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00071594
CAS Number: 6138-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR