5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
A146608-100G
A146608-10G
A146608-1MG
A146608-250MG
A146608-500G
A146608-1G
A146608-25G
A146608-5G
103803-688EA
9
USD
103803-688
103841-782
76783-186
76783-188
76783-190
76783-192
76783-194
77299-538
77299-540
77317-116
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) 98%
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
Precaución: for Research Use Only
Formula:
C₁₄H₈N₂O₈S₂ MW: 396.36 g/mol Boiling Pt: 671.9 °C (760 mm Hg) Melting Pt: 243…245 °C Flash Pt: 360.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007140 CAS Number: 69-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...