(R)-(-)-tert-Leucinol 97%

Supplier: Ambeed
Warning

A492983-1G A492983-5G A492983-10G A492983-250MG
103828-232EA 12 USD
103828-232 103839-308 76843-268 76843-270 77143-970 77143-972
(R)-(-)-tert-Leucinol 97%
(R)-(-)-tert-Leucinol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
Boiling Pt: 70 °C (760 mm Hg)
Melting Pt: 30…33 °C
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01076574
CAS Number: 112245-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR