D-Ribofuranose, 1-acetate 2,3,5-tribenzoate 98%

Supplier: Ambeed
Warning

Synonyms: (3R,4R,5R)-2-Acetoxy-5-((benzoyloxy)methyl)tetrahydrofuran-3,4-diyl dibenzoate, 1-O-Acetyl-2,3,5-tri-O-benzoyl-D-ribofuranose

A278389-100G A278389-10G A278389-25G A278389-5G
103853-354EA 7 USD
103853-354 77014-546 77014-548 77093-270 77093-272
D-Ribofuranose, 1-acetate 2,3,5-tribenzoate 98%
D-Ribofuranose, 1-acetate 2,3,5-tribenzoate

Precaución: for Research Use Only

Formula: C₂₈H₂₄O₉
MW: 504.48 g/mol
Boiling Pt: 621.0±55.0 °C (760 mm Hg)
Melting Pt: 126…127 °C
Flash Pt: 264.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00067634
CAS Number: 14215-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR