(R)-(-)-Prolinol 98%

Supplier: Ambeed
Warning

A209217-100G A209217-10G A209217-500G A209217-1G A209217-25G A209217-5G
103817-922EA 9 USD
103817-922 103848-610 76986-652 76986-654 76986-656 77303-088 77303-090 77303-092
(R)-(-)-Prolinol 98%
(R)-(-)-Prolinol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
Boiling Pt: 211 °C (760 mm Hg)
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064321
CAS Number: 68832-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR