(R)-(-)-β-Aminophenethyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: D-Phenylglycinol, (R)-(-)-2-Phenylglycinol

A276015-100G A276015-10G A276015-1G A276015-500G A276015-1000G A276015-25G A276015-5G
103809-912EA 177 USD
103809-912 103853-216 76772-220 76772-222 76772-224 76772-226 77093-074 77093-076 77305-224
(R)-(-)-β-Aminophenethyl alcohol 98%
(R)-(-)-β-Aminophenethyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Flash Pt: 125.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008062
CAS Number: 56613-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR