D-(-)-Penicillamine 98%

Supplier: Ambeed
Warning

A101389-100MG A101389-250MG A101389-25G A101389-50MG A101389-5G A101389-1G A101389-100G A101389-500G
103795-950EA 12 USD
103795-950 103821-634 76970-166 76970-168 76970-170 76970-172 77282-716 77297-084 77548-908 77670-464
D-(-)-Penicillamine 98%
D-(-)-Penicillamine

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂S
Boiling Pt: 251.8±35.0 °C (760 mm Hg)
Melting Pt: 210 °C
Flash Pt: 106.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064302
CAS Number: 52-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR