Docosahexaenoic acid 95%

Supplier: Ambeed
Warning

A267106-100MG A267106-10G A267106-250MG A267106-25G A267106-1G A267106-5G
103799-542EA 36 USD
103799-542 103812-838 76692-778 76692-780 76692-782 76692-784 77304-970 77626-398
Docosahexaenoic acid 95%
Docosahexaenoic acid

Precaución: for Research Use Only

Formula: C₂₂H₃₂O₂
MW: 328.488 g/mol
Boiling Pt: 446.732 °C (760 mm Hg)
Melting Pt: 44 °C (760 mm Hg)
Flash Pt: 343.387 °C
Storage Temperature: Freezer
MDL Number: MFCD00065722
CAS Number: 6217-54-5
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR