Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7-triacetate 98%

Supplier: Ambeed
Warning

Synonyms: 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic Acid Tri-tert-butyl Ester, Tri-tert-butyl 2,2',2''-(1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate

A160100-100G A160100-10G A160100-25G A160100-1G A160100-5G
103805-332EA 23 USD
103805-332 103843-562 77150-828 77150-830 77150-832 77319-480 77319-482
Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7-triacetate 98%
Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7-triacetate

Precaución: for Research Use Only

Formula: C₂₆H₅₀N₄O₆
MW: 514.71 g/mol
Boiling Pt: 561.6 °C (760 mm Hg)
Melting Pt: 181…183 °C
Flash Pt: 293.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05865261
CAS Number: 122555-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR