(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2:5,6-Di-O-isopropylidene-D-mannitol, (1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol, (1S,2S)-1,2-BIS((R)-2,2-DIMETHYL-1,3-DIOXOLAN-4-YL)ETHANE-1,2-DIOL
A158555-5G
A158555-100G
A158555-500G
103808-866EA
345
USD
103808-866
103843-352
77300-338
77592-930
77671-246
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol 95%
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
Precaución: for Research Use Only
Formula:
C₁₂H₂₂O₆ MW: 262.302 g/mol Boiling Pt: 382 °C (760 mm Hg) Melting Pt: 120…122 °C Flash Pt: 184.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003211 CAS Number: 1707-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...