DL-Homocystine 95%

Supplier: Ambeed
Warning

Synonyms: H-DL-HoCystine-OH, rel-(2S,2'S)-4,4'-Disulphanediylbis(2-aminobutanoic acid)

A263842-10G A263842-500G
103852-532EA 8 USD
103852-532 77153-526 77153-528
DL-Homocystine 95%
DL-Homocystine

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₄S₂
MW: 268.35 g/mol
Boiling Pt: 507.6±50.0 °C (760 mm Hg)
Melting Pt: >300 °C
Flash Pt: 260.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00069587
CAS Number: 870-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR