DL-Histidine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(1H-imidazol-4-yl)propanoic acid, H-DL-His-OH

A890579-100G A890579-10G
103841-332EA 13 USD
103841-332 77106-128 77106-130
DL-Histidine 95%
DL-Histidine

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Boiling Pt: 458.9 °C (760 mm Hg)
Melting Pt: 273
Storage Temperature: Ambient
MDL Number: MFCD00005208
CAS Number: 4998-57-6
Merck Index: 12,04758

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR