DL-2-Allylglycine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
DL-2-Amino-4-pentenoic acid, DL-2-Allylglycine, 2-Amino-4-pentenoic Acid, 2-Aminopent-4-enoic acid
A190290-1G
A190290-5G
A190290-10G
A190290-25G
A190290-100G
A190290-250MG
103816-280EA
17
USD
103816-280
103846-882
77084-668
77084-670
77164-256
77302-220
77683-495
77683-496
DL-2-Allylglycine 95%
DL-2-Allylglycine
Precaución: For research use only.
Formula:
C₅H₉NO₂ MW: 115.13 g/mol Melting Pt: 258…260 °C Storage Temperature: Freezer |
MDL Number:
MFCD00063103 CAS Number: 7685-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...