2-Amino-2-(4-bromophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: DL-2-Amino-2-(4-bromophenyl)acetic acid

A341603-25G A341603-5G A341603-100G A341603-10G A341603-250MG A341603-1G
103825-838EA 36 USD
103825-838 103836-812 76839-842 76839-844 77239-882 77239-884 77239-886 77306-540
2-Amino-2-(4-bromophenyl)acetic acid 97%
2-Amino-2-(4-bromophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
Melting Pt: 273…274 °C
Flash Pt: 173.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00049327
CAS Number: 71079-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR