Di-tert-butylphenylphosphonium tetrafluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Di-tert-butyl(phenyl)phosphonium tetrafluoroborate

A331025-1G A331025-25G A331025-5G
103836-532EA 37 USD
103836-532 76803-058 76803-060 76839-514
Di-tert-butylphenylphosphonium tetrafluoroborate 98%
Di-tert-butylphenylphosphonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₄H₂₄BF₄P
MW: 310.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08704553
CAS Number: 612088-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR