2-(Di-tert-butylphosphino)-2'-methylbiphenyl 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Di-t-butylphosphino-2'-methyl)-1,1'-biphenyl, t-BuMePhos, Di-tert-butyl(2'-methyl-[1,1'-biphenyl]-2-yl)phosphine
A100227-100G
A100227-10G
A100227-250MG
A100227-25G
A100227-1G
A100227-5G
103795-306EA
14
USD
103795-306
103821-018
77245-066
77245-068
77245-070
77245-072
77296-600
77296-602
2-(Di-tert-butylphosphino)-2'-methylbiphenyl 97%
2-(Di-tert-butylphosphino)-2'-methylbiphenyl
Precaución: for Research Use Only
Formula:
C₂₁H₂₉P MW: 312.437 g/mol Boiling Pt: 408 °C (760 mm Hg) Melting Pt: 105…109 °C Flash Pt: 211.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD03453047 CAS Number: 255837-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...