Di-tert-butyl iminodicarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Di-(tert-butyl) imidodicarbonate

A301919-1000G A301919-100G A301919-10G A301919-500G A301919-25G A301919-1G A301919-5G
103814-318EA 5 USD
103814-318 103835-474 76693-420 76693-422 76693-424 76693-426 76838-166 77305-840 77305-842
Di-tert-butyl iminodicarboxylate 98%
Di-tert-butyl iminodicarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
Boiling Pt: 273.8 °C (760 mm Hg)
Melting Pt: 114…117 °C
Flash Pt: 119.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00043309
CAS Number: 51779-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR