Di-tert-butyl hydrazodicarboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Di-tert-butyl hydrazine-1,2-dicarboxylate, Di-tert-butyl Hydrazodiformate, Di-tert-butyl 1,2-Hydrazinedicarboxylate
A297341-100G
A297341-10G
A297341-500G
A297341-25G
A297341-5G
A297341-1G
103809-994EA
307
USD
103809-994
103814-130
103835-272
76802-494
76802-496
76802-498
76837-958
76837-960
77305-704
Di-tert-butyl hydrazodicarboxylate 98%
Di-tert-butyl hydrazodicarboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₄ MW: 232.28 g/mol Boiling Pt: 294.6±9.0 °C (760 mm Hg) Melting Pt: 123…126 °C Flash Pt: 132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015000 CAS Number: 16466-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...