Di-tert-butyl azodicarboxylate 97%

Supplier: Ambeed
Danger

Synonyms: Di-tert-butyl diazene-1,2-dicarboxylate

A257862-10G A257862-500G A257862-25G A257862-5G
103809-830EA 394 USD
103809-830 103852-210 76692-660 76692-662 76692-664 77091-736 77304-720
Di-tert-butyl azodicarboxylate 97%
Di-tert-butyl azodicarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₄
Melting Pt: 89…92 °C
Flash Pt: 107.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00015001
CAS Number: 870-50-8
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR