Di-tert-butyl Iminodiacetate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Iminodiacetic acid Di-tert-butyl Ester, Di-tert-butyl 2,2'-azanediyldiacetate
A157234-10G
A157234-250MG
A157234-25G
A157234-5G
103804-980EA
36
USD
103804-980
103843-180
76798-830
76798-832
76798-834
77300-242
Di-tert-butyl Iminodiacetate 97%
Di-tert-butyl Iminodiacetate
Precaución: for Research Use Only
Formula:
C₁₂H₂₃NO₄ Boiling Pt: 298 °C (760 mm Hg) Flash Pt: 134 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01863672 CAS Number: 85916-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...