1,7-Bis-boc-1,4,7-triazaheptane 97%

Supplier: Ambeed
Warning

A108361-100MG A108361-10G A108361-250MG A108361-25G A108361-1G A108361-5G A108361-100G
103797-334EA 74 USD
103797-334 103823-114 76869-558 76869-560 76869-562 76869-564 77309-958 77309-960 77565-910
1,7-Bis-boc-1,4,7-triazaheptane 97%
1,7-Bis-boc-1,4,7-triazaheptane

Precaución: For research use only.

Formula: C₁₄H₂₉N₃O₄
Storage Temperature: Refrigerator
MDL Number: MFCD11226825
CAS Number: 117499-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR