Disulfiram 97%

Supplier: Ambeed
Danger

A912727-1MG A912727-25G A912727-100G A912727-500G A912727-1000G
103841-382EA 16 USD
103841-382 76698-676 77308-512 77603-276 77627-738 77878-734
Disulfiram 97%
Disulphiram

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂S₄
Storage Temperature: Ambient
MDL Number: MFCD00009048
CAS Number: 97-77-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR