Disodium UDP-glucose 97%

Supplier: Ambeed
Warning

Synonyms: ((2R,3S,4R,5R)-5-(2,4-Dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl ((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)diphosphate sodium, UDPG, UDP-α-D-Glucose disodium salt, UDP-Glucose, Uridine 5′-diphosphoglucose disodium salt, Uridine-5'-diphosphoglucose, Uridine-5′-diphosphoglucose disodium salt

A203590-250MG A203590-1G
103817-472EA 162 USD
103817-472 103848-130 77139-404 77302-860
Disodium UDP-glucose 97%
Disodium UDP-glucose

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂Na₂O₁₇P₂
MW: 610.26 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077895
CAS Number: 28053-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR