Rhodium(II) acetate dimer 98%

Supplier: Ambeed
Warning

Synonyms: Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)

A350806-100MG A350806-10G A350806-250MG
103826-042EA 487 USD
103826-042 103837-038 76724-458 76724-460 76724-462
Rhodium(II) acetate dimer 98%
Rhodium (II) acetate dimer

Precaución: for Research Use Only

Formula: C₈H₁₂O₈Rh₂
MW: 441.987 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003538
CAS Number: 15956-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR