(-)-Di-p-toluoyl-L-tartaric acid 98%

Supplier: Ambeed
Warning

Synonyms: Di-p-toluoyl-L-tartaric acid, (-)-O,O'-Di-p-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric Acid

A330098-25G A330098-100G A330098-500G
103810-148EA 57 USD
103810-148 76839-482 77290-418 77290-420
(-)-Di-p-toluoyl-L-tartaric acid 98%
(-)-Di-p-toluoyl-L-tartaric acid

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₈
Boiling Pt: 626.5 °C (760 mm Hg)
Melting Pt: 169…171 °C
Flash Pt: 223.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00064440
CAS Number: 32634-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR