Benzhydryl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: Diphenylcarbinol, Benzhydrol, Diphenylmethanol

A260557-100G A260557-25G A260557-500G A260557-1000G
103809-854EA 33 USD
103809-854 76874-846 76874-848 76874-850 77091-968
Benzhydryl alcohol 98%
Benzhydryl alcohol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O
MW: 184.24 g/mol
Boiling Pt: 297…298 °C (760 mm Hg)
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00004488
CAS Number: 91-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR