Diphenyliodonium trifluoromethanesulfonate 98%

Supplier: Ambeed
Danger

A100258-10G A100258-250MG A100258-25G A100258-1G A100258-5G
103795-326EA 12 USD
103795-326 103821-036 76796-220 76796-222 76796-224 77296-618 77296-620
Diphenyliodonium trifluoromethanesulfonate 98%
Diphenyliodonium trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₃IO₃S
Melting Pt: 180…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00191356
CAS Number: 66003-76-7
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR