Diphenyliodonium chloride 97%
Supplier: Ambeed
|
Danger
|
A139147-1G
A139147-25G
A139147-5G
103802-704EA
13
USD
103802-704
77315-780
77315-782
77315-784
Diphenyliodonium chloride 97%
Diphenyliodonium chloride
Precaución: for Research Use Only
Formula:
C₁₂H₁₀ClI MW: 316.57 g/mol Melting Pt: 227…235 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011909 CAS Number: 1483-72-3 UN: 2811 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...