Diphenylcyclopropenone 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one

A632068-250MG A632068-1G A632068-25G A632068-5G
103840-280EA 71 USD
103840-280 76806-226 76844-452 76844-454 76844-456
Diphenylcyclopropenone 98%
Diphenylcyclopropenone

Precaución: for Research Use Only

Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00001311
CAS Number: 886-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR