Diphenylacetylene 98%

Supplier: Ambeed
Warning

A360795-5G A360795-100G A360795-500G A360795-25G
103837-282EA 14 USD
103837-282 76840-494 77106-914 77106-916 77306-718
Diphenylacetylene 98%
Diphenylacetylene

Precaución: for Research Use Only

Formula: C₁₄H₁₀
MW: 178.23 g/mol
Melting Pt: 58…61 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00004786
CAS Number: 501-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR