Diphenyl(2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphine 97%

Supplier: Ambeed
Warning

A121699-250MG A121699-25G A121699-1G A121699-5G
103800-110EA 8 USD
103800-110 76810-508 76810-510 77312-236 77312-238
Diphenyl(2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphine 97%
Diphenyl(2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphine

Precaución: for Research Use Only

Formula: C₃₃H₃₇P
MW: 464.62 g/mol
Boiling Pt: 545.3±50.0 °C (760 mm Hg)
Flash Pt: 301.7 °C
Storage Temperature: Ambient
MDL Number: MFCD26961066
CAS Number: 819867-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR