Diphenyl phosphate 97%

Supplier: Ambeed
Warning

Synonyms: Diphenyl hydrogen phosphate

A114619-25G A114619-500G A114619-1000G A114619-5G A114619-100G
103808-126EA 147 USD
103808-126 103824-274 76797-032 76797-034 77311-094 77311-096 77549-244
Diphenyl phosphate 97%
Diphenyl phosphate

Precaución: for Research Use Only

Formula: C₁₂H₁₁O₄P
MW: 250.19 g/mol
Boiling Pt: 377.7 °C (760 mm Hg)
Melting Pt: 63…70 °C
Flash Pt: 182.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00003033
CAS Number: 838-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR