Diosmetin 96%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-chromen-4-one, 5,7,3'-Trihydroxy-4'-methoxyflavone

A124051-10G A124051-250MG A124051-1G A124051-100MG A124051-50MG A124051-100G A124051-1MG A124051-25G
103800-478EA 31 USD
103800-478 76797-574 76797-576 77312-606 77592-416 77599-804 77624-034 77624-036 77624-038
Diosmetin 96%
Diosmetin

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₆
MW: 300.27 g/mol
Boiling Pt: 576.7 °C (760 mm Hg)
Melting Pt: 256…258 °C
Flash Pt: 220.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017425
CAS Number: 520-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR