Dimethyl 3,5-Pyridinedicarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl pyridine-3,5-dicarboxylate, 3,5-Pyridinedicarboxylic acid Dimethyl ester

A290103-100G A290103-250MG A290103-1G A290103-10G
103813-860EA 6 USD
103813-860 103835-000 76802-364 76802-366 76837-580 77593-776
Dimethyl 3,5-Pyridinedicarboxylate 98%
Dimethyl 3,5-Pyridinedicarboxylate

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Boiling Pt: 267.6 °C (760 mm Hg)
Melting Pt: >80 °C
Flash Pt: 115.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00518827
CAS Number: 4591-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR