Dimethyl-2,3-pyridinedicarboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dimethyl pyridine-2,3-dicarboxylate
A159998-100G
A159998-10G
A159998-1G
A159998-500G
A159998-25G
A159998-5G
103843-542EA
9
USD
103843-542
76689-782
76689-784
76689-786
76689-788
77319-446
77319-448
Dimethyl-2,3-pyridinedicarboxylate 98%
Dimethyl 2,3-Pyridinedicarboxylate
Precaución: for Research Use Only
Formula:
C₉H₉NO₄ MW: 195.17 g/mol Boiling Pt: 262.3 °C (760 mm Hg) Melting Pt: 56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00186549 CAS Number: 605-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...