Dimethyl glutaconate (mixture of cis and trans isomers) 95%

Supplier: Ambeed
Warning

Synonyms: Dimethyl pent-2-enedioate

A157657-100G A157657-10G A157657-25G A157657-5G A157657-1G
103805-032EA 11 USD
103805-032 103843-234 76798-856 76798-858 77300-278 77300-280 77319-070
Dimethyl glutaconate (mixture of cis and trans isomers) 95%
Dimethyl glutaconate (mixture of cis and trans isomers)

Precaución: for Research Use Only

Formula: C₇H₁₀O₄
Boiling Pt: 113…114 °C at 150 mmHg
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00066240
CAS Number: 5164-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR