Dimethyl furan-2,5-dicarboxylate 98%

Supplier: Ambeed
Warning

A238586-100G A238586-10G A238586-1G A238586-5G A238586-500G A238586-1000G
103811-236EA 8 USD
103811-236 103851-020 77031-224 77031-226 77090-140 77090-142 77550-538 77872-199
Dimethyl furan-2,5-dicarboxylate 98%
Dimethyl furan-2,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₈H₈O₅
Boiling Pt: 270.9 °C (760 mm Hg)
Flash Pt: 117.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00092317
CAS Number: 4282-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR