Dimethyl cyclohexane-1,3-dicarboxylate 95%

Supplier: Ambeed
Warning

A115856-100MG A115856-1G
103798-636EA 16 USD
103798-636 103824-498 76687-820 77311-324
Dimethyl cyclohexane-1,3-dicarboxylate 95%
Dimethyl cyclohexane-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
Boiling Pt: 258.1 °C (760 mm Hg)
Flash Pt: 120.7 °C
Storage Temperature: Ambient
MDL Number: MFCD07779254
CAS Number: 62638-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR