Dimethyl-7,7'-dimethoxy[4,4'-bibenzo[d][1,3]dioxole]5,5'-dicarboxylate 99+%

Supplier: Ambeed
Warning

A100986-100G A100986-10G A100986-25G A100986-1G A100986-5G
103795-736EA 7 USD
103795-736 103821-428 77107-186 77107-188 77107-190 77282-632 77282-634
Dimethyl-7,7'-dimethoxy[4,4'-bibenzo[d][1,3]dioxole]5,5'-dicarboxylate 99+%
Dimethyl-7,7'-dimethoxy[4,4'-bibenzo[d][1,3]dioxole]5,5'-dicarboxylate

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₁₀
Boiling Pt: 606.9 °C (760 mm Hg)
Flash Pt: 265.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01751431
CAS Number: 73536-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR