Dimethyl-5-iodoisophthalate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Iodoisophthalic acid dimethyl ester, Dimethyl 5-iodoisophthalate

A317955-100G A317955-10G A317955-250MG A317955-1G A317955-25G A317955-5G
103825-158EA 6 USD
103825-158 103836-088 76772-872 76772-874 76772-876 77306-162 77306-164 77306-166
Dimethyl-5-iodoisophthalate 98%
Dimethyl-5-iodoisophthalate

Precaución: for Research Use Only

Formula: C₁₀H₉IO₄
MW: 320.08
Boiling Pt: 367.7 °C (760 mm Hg)
Melting Pt: 105 °C
Flash Pt: 176.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00079753
CAS Number: 51839-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR